missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-fluorobenzene, 98%

CAS: 766-98-3 | C8H5F | 120.13 g/mol

$121.39 - $121.39

Chemical Identifiers

CAS 766-98-3
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.13
MDL Number MFCD00168823
InChI Key QXSWHQGIEKUBAS-UHFFFAOYSA-N
Synonym 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
PubChem CID 522627
IUPAC Name 1-ethynyl-4-fluorobenzene
SMILES C#CC1=CC=C(C=C1)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371370010
SDS View Documents
Thermo Scientific Chemicals
371370010
1 g Glass bottle
Each for $121.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

766-98-3
120.13
QXSWHQGIEKUBAS-UHFFFAOYSA-N
522627
C#CC1=CC=C(C=C1)F
C8H5F
MFCD00168823
4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
1-ethynyl-4-fluorobenzene

Specifications

766-98-3
100.0
Colorless to Yellow
55.0°C to 56.0°C (40.0 mmHg)
Authentic
Glass bottle
1.5150 to 1.5170
MFCD00168823
1.048
QXSWHQGIEKUBAS-UHFFFAOYSA-N
1-ethynyl-4-fluorobenzene
522627
98%
1-Ethynyl-4-fluorobenzene, 98%
97.5
27.0°C to 28.0°C
1.0480g/mL
29°C
97.5% min. (GC)
C8H5F
FC6H4C≡CH
1 g
4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
C#CC1=CC=C(C=C1)F
120.13
120.13
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only