missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynyl-3,5-dimethoxybenzene 98.0+%, TCI America™
Supplier: TCI America E11751G
Specifications
| 1-Ethynyl-3,5-dimethoxybenzene | |
| 46°C | |
| C10H10O2 | |
| 1 g | |
| HUSBBWQIJMRKLI-UHFFFAOYSA-N | |
| 1-ethynyl-3,5-dimethoxybenzene | |
| 4661321 | |
| ≥98.0% (GC) |
| 171290-52-1 | |
| White-Yellow | |
| MFCD03839985 | |
| 3,5-Dimethoxyphenylacetylene | |
| COC1=CC(=CC(OC)=C1)C#C | |
| 162.19 | |
| 162.19 | |
| Crystalline Powder |
Chemical Identifiers
| 171290-52-1 | |
| 162.19 | |
| HUSBBWQIJMRKLI-UHFFFAOYSA-N | |
| 4661321 | |
| COC1=CC(=CC(OC)=C1)C#C |
| C10H10O2 | |
| MFCD03839985 | |
| 3,5-Dimethoxyphenylacetylene | |
| 1-ethynyl-3,5-dimethoxybenzene |
Safety and Handling
TSCA : No