accessibility menu, dialog, popup

UserName

CH3OHCOCH3

CAS RN 171290-52-1

IUPAC Name: 1-ethynyl-3,5-dimethoxybenzene
Synonyms: 1-ethynyl-3,5-dimethoxybenzene
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: HUSBBWQIJMRKLI-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)