missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-3,5-dimethoxybenzene 98.0+%, TCI America™

Catalog No. E1175200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1175200MG 200 mg
E11751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1175200MG Supplier TCI America Supplier No. E1175200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 171290-52-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD03839985
InChI Key HUSBBWQIJMRKLI-UHFFFAOYSA-N
Synonym 3,5-Dimethoxyphenylacetylene
PubChem CID 4661321
IUPAC Name 1-ethynyl-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(OC)=C1)C#C

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 200 mg
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethynyl-3,5-dimethoxybenzene