missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpyridinium Chloride 98.0+%, TCI America™

Catalog No. p-7129057
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05445G 5 g
E054425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05445G Supplier TCI America Supplier No. E05445G

Chemical Identifiers

CAS 2294-38-4
Molecular Formula C7H10ClN
Molecular Weight (g/mol) 143.614
MDL Number MFCD03093293
InChI Key AMFMJCAPWCXUEI-UHFFFAOYSA-M
Synonym 1-ethylpyridinium chloride, ethylpyridinium chloride, 1-ethylpyridin-1-ium chloride, unii-38bql5563g, pyridinium, 1-ethyl-, chloride, n-ethylpyridinium chloride, pyridinium, 1-ethyl-, chloride 1:1, 1-ethylpyridiniumchloride, pyridine ethochloride, ethylpyridiniumchloride
PubChem CID 75299
IUPAC Name 1-ethylpyridin-1-ium;chloride
SMILES CC[N+]1=CC=CC=C1.[Cl-]

Specifications

Color Blue-White
Quantity 5 g
Formula Weight 143.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Ethylpyridinium Chloride