accessibility menu, dialog, popup

UserName

ClH3CN

CAS RN 2294-38-4

IUPAC Name: 1-ethylpyridin-1-ium chloride
Synonyms: 1-ethylpyridin-1-ium chloride
Molecular Weight (g/mol): 143.61
Molecular Formula: C7H10ClN
InChi Key: AMFMJCAPWCXUEI-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+]1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results