missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyridinium Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Blue-White |
| Quantity | 5 g |
| Formula Weight | 143.61 |
| Percent Purity | ≥98.0% (T) |
| Grade | Research |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 1-Ethylpyridinium Chloride |
Safety and Handling
| TSCA | Yes |