missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyrazole 98.0+%, TCI America™
$44.73 - $139.81
Chemical Identifiers
| CAS | 2817-71-2 |
|---|---|
| Molecular Formula | C5H8N2 |
| Molecular Weight (g/mol) | 96.133 |
| MDL Number | MFCD03444367 |
| InChI Key | FLNMQGISZVYIIK-UHFFFAOYSA-N |
| PubChem CID | 498446 |
| IUPAC Name | 1-ethylpyrazole |
| SMILES | CCN1C=CC=N1 |
Chemical Identifiers
| 2817-71-2 | |
| 96.133 | |
| FLNMQGISZVYIIK-UHFFFAOYSA-N | |
| 1-ethylpyrazole |
| C5H8N2 | |
| MFCD03444367 | |
| 498446 | |
| CCN1C=CC=N1 |
Specifications
| 2817-71-2 | |
| 137°C | |
| MFCD03444367 | |
| 1993 | |
| CCN1C=CC=N1 | |
| 96.133 | |
| 96.13 | |
| Liquid |
| Yellow | |
| C5H8N2 | |
| 5 mL | |
| FLNMQGISZVYIIK-UHFFFAOYSA-N | |
| 1-ethylpyrazole | |
| 498446 | |
| ≥98.0% (GC,T) | |
| 1-Ethylpyrazole |
Safety and Handling
TSCA : No