accessibility menu, dialog, popup

UserName

CAS RN 2817-71-2

H3CNN

CAS RN 2817-71-2

IUPAC Name: 1-ethyl-1H-pyrazole
Synonyms: 1-ethylpyrazole
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: FLNMQGISZVYIIK-UHFFFAOYSA-N
SMILES: CCN1C=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results