missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylnaphthalene 97.0+%, TCI America™

Catalog No. E014610ML
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E014610ML 10 mL
E014625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. E014610ML missing translation for 'mfr' TCI America missing translation for 'supplierNo' E014610ML

Chemical Identifiers

CAS 1127-76-0
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004049
InChI Key ZMXIYERNXPIYFR-UHFFFAOYSA-N
Synonym naphthalene, 1-ethyl, ethylnaphthalene, naphthalene, ethyl, alpha-ethylnaphthalene, .alpha.-ethylnaphthalene, 1-ethyl-naphthalene, 1-ethylphthalene, 1-ethyl-naphthalen, 1-ethyl naphthalene, acmc-1boes
PubChem CID 14315
IUPAC Name 1-ethylnaphthalene
SMILES CCC1=C2C=CC=CC2=CC=C1

Specifications

Color Colorless
Boiling Point 260°C
Quantity 10 mL
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethylnaphthalene
RTECSNumber QJ6950000
TSCA No