accessibility menu, dialog, popup

UserName

H3C

CAS RN 1127-76-0

IUPAC Name: 1-ethylnaphthalene
Synonyms: 1-ethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: ZMXIYERNXPIYFR-UHFFFAOYSA-N
SMILES: CCC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)