missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylnaphthalene, 98%

Catalog No. AAH5641314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5641314 25 g
AAH5641306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5641314 Supplier Thermo Scientific Chemicals Supplier No. H5641314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1127-76-0 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1127-76-0
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004049
InChI Key ZMXIYERNXPIYFR-UHFFFAOYSA-N
Synonym naphthalene, 1-ethyl, ethylnaphthalene, naphthalene, ethyl, alpha-ethylnaphthalene, .alpha.-ethylnaphthalene, 1-ethyl-naphthalene, 1-ethylphthalene, 1-ethyl-naphthalen, 1-ethyl naphthalene, acmc-1boes
PubChem CID 14315
IUPAC Name 1-ethylnaphthalene
SMILES CCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point -15°C
Color Colorless to Yellow
Density 1.008 g/mL
Boiling Point 258°C to 260°C
Flash Point 111°C (232°F)
Quantity 25 g
Beilstein 1854417
Formula Weight 156.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethylnaphthalene
Show More Show Less
EINECSNumber 214-432-4
RTECSNumber QJ6950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only