missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylcyclopentanol 96.0+%, TCI America™

Catalog No. p-7129147
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07825G 5 g
E078225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07825G Supplier TCI America Supplier No. E07825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1462-96-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00019281
InChI Key LPCWIFPJLFCXRS-UHFFFAOYSA-N
Synonym 1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, ksc181q7f, 1-ethyl-1-hydroxycyclopentane
PubChem CID 137018
IUPAC Name 1-ethylcyclopentan-1-ol
SMILES CCC1(CCCC1)O

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 153°C
Quantity 5 g
UN Number 1987
Formula Weight 114.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethylcyclopentanol