missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylamino-2-propanol 97.0+%, TCI America™

Catalog No. E12445ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12445ML 5 mL
E124425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12445ML Supplier TCI America Supplier No. E12445ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40171-86-6
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00082193
InChI Key UCYJVNBJCIZMTJ-UHFFFAOYNA-N
PubChem CID 3016164
IUPAC Name 1-(ethylamino)propan-2-ol
SMILES CCNCC(C)O

Specifications

Melting Point 14°C
Color Colorless
Boiling Point 160°C
Quantity 5 mL
UN Number 2735
Formula Weight 103.17
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Ethylamino-2-propanol
EINECSNumber (9)-1916
TSCA No