accessibility menu, dialog, popup

UserName

H3CNHCH3OH

CAS RN 40171-86-6

IUPAC Name: 1-(ethylamino)propan-2-ol
Synonyms: 1-(ethylamino)propan-2-ol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: UCYJVNBJCIZMTJ-UHFFFAOYNA-N
SMILES: CCNCC(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results