missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-methylpyridinium Bromide 98.0+%, TCI America™

Catalog No. E12725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12725G 5 g
E127225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12725G Supplier TCI America Supplier No. E12725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32353-49-4
Molecular Formula C8H12BrN
Molecular Weight (g/mol) 202.095
MDL Number MFCD18384825
InChI Key YZBRZOWPIBQUEJ-UHFFFAOYSA-M
PubChem CID 14358228
IUPAC Name 1-ethyl-4-methylpyridin-1-ium;bromide
SMILES CC[N+]1=CC=C(C=C1)C.[Br-]

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 202.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-4-methylpyridinium Bromide