accessibility menu, dialog, popup

UserName

CAS RN 32353-49-4

H3CNCH3Br

CAS RN 32353-49-4

IUPAC Name: 1-ethyl-4-methylpyridin-1-ium bromide
Synonyms: 1-ethyl-4-methylpyridin-1-ium bromide
Molecular Weight (g/mol): 202.1
Molecular Formula: C8H12BrN
InChi Key: YZBRZOWPIBQUEJ-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results