missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-iodobenzene, 98%

Catalog No. p-7021696
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1413809 10 g
AAA1413818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1413809 Supplier Thermo Scientific Chemicals Supplier No. A1413809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25309-64-2 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25309-64-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00060663
InChI Key OOLSRHZMXAYDFB-UHFFFAOYSA-N
Synonym 4-ethyliodobenzene, benzene, 1-ethyl-4-iodo, pubchem3877, 1-iodo-4-ethylbenzene, acmc-1chvo, p-ethyliodobenzene, 1-ethyl-4-iodo-benzene, #, 1-ethyl-4-iodanyl-benzene
PubChem CID 141189
IUPAC Name 1-ethyl-4-iodobenzene
SMILES CCC1=CC=C(C=C1)I

Specifications

Melting Point -17°C
Density 1.6
Boiling Point 112°C to 113°C (20 mmHg)
Flash Point 107°C (224°F)
Odor Mild
Refractive Index 1.591
Quantity 10 g
Beilstein 2076973
Sensitivity Light sensitive
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 1-Ethyl-4-iodobenzene
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only