missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-eth-1-ynylbenzene, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09906EA

Quantity 10g

 View more versions of this product

Catalog No. BTB09906EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

40307-11-7
130.19
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
142425
CCC1=CC=C(C=C1)C#C
C10H10
MFCD00173887
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
1-ethyl-4-ethynylbenzene

Specifications

1-Ethyl-4-eth-1-ynylbenzene
96.5
0.93g/mL
Amber glass bottle
HC2CC6H4CH2CH3
MFCD00173887
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
1-ethyl-4-ethynylbenzene
142425
97%
40307-11-7
100.0
97%
C10H10
10g
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
CCC1=CC=C(C=C1)C#C
130.19
130.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents