missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene 98.0+%, TCI America™

Catalog No. E08061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08061G 1 g
E08065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08061G Supplier TCI America Supplier No. E08061G

Chemical Identifiers

CAS 63221-88-5
Molecular Formula C17H16O
Molecular Weight (g/mol) 236.31
MDL Number MFCD00807428
InChI Key YWDALJMLXXWXFP-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-methoxyphenyl ethynyl benzene, 1-4-ethylphenyl-2-4-methoxyphenyl acetylene, 4-4-ethylphenyl ethynyl anisole, 1-ethyl-4-2-4-methoxyphenyl ethynyl benzene, benzene,1-ethyl-4-2-4-methoxyphenyl ethynyl, 1-2-4-ethylphenyl ethynyl-4-methoxybenzene, 4-methoxy-4'-ethyltolane, acmc-209ne1
PubChem CID 3952083
IUPAC Name 1-ethyl-4-[2-(4-methoxyphenyl)ethynyl]benzene
SMILES CCC1=CC=C(C=C1)C#CC1=CC=C(OC)C=C1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 153°C
Quantity 1 g
Formula Weight 236.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene