missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylpyrazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. E12281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12281G 1 g
E12285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12281G Supplier TCI America Supplier No. E12281G

Chemical Identifiers

CAS 50920-65-5
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00156147
InChI Key VFMGOJUUTAPPDA-UHFFFAOYSA-N
Synonym 1-ethyl-3-methyl-1h-pyrazole-5-carboxylic acid, 1-ethyl-3-methylpyrazole-5-carboxylic acid, 2-ethyl-5-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-ethyl-3-methyl, maybridge1_008579, acmc-20aovb, 1-ethyl-3-methyl-1h-pyrazole-5-carboxylicacid, 1-ethyl-3-methylpyrazole-5-carboxylic, 1h-pyrazole-5-carboxylicacid, 1-ethyl-3-methyl
PubChem CID 2743794
IUPAC Name 2-ethyl-5-methylpyrazole-3-carboxylic acid
SMILES CCN1C(=CC(=N1)C)C(=O)O

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylpyrazole-5-carboxylic Acid