missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Diethyl Phosphate 96.0+%, TCI America™
Supplier: TCI America E130925G
Specifications
| 1-Ethyl-3-methylimidazolium Diethyl Phosphate | |
| C10H21N2O4P | |
| 25 g | |
| 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc | |
| CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC | |
| 264.26 | |
| 264.26 | |
| Liquid |
| 848641-69-0 | |
| MFCD09953486 | |
| 3265 | |
| HQWOEDCLDNFWEV-UHFFFAOYSA-M | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium diethyl phosphate | |
| 53316406 | |
| ≥96.0% (HPLC,T) |
Chemical Identifiers
| 848641-69-0 | |
| 264.26 | |
| HQWOEDCLDNFWEV-UHFFFAOYSA-M | |
| 53316406 | |
| CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC |
| C10H21N2O4P | |
| MFCD09953486 | |
| 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium diethyl phosphate |
Safety and Handling
TSCA : No