missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Diethyl Phosphate 96.0+%, TCI America™

Catalog No. p-7129428
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13095G 5 g
E130925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13095G Supplier TCI America Supplier No. E13095G
Only null left

Chemical Identifiers

CAS 848641-69-0
Molecular Formula C10H21N2O4P
Molecular Weight (g/mol) 264.26
MDL Number MFCD09953486
InChI Key HQWOEDCLDNFWEV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
PubChem CID 53316406
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium diethyl phosphate
SMILES CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC

Specifications

Quantity 5 g
UN Number 3265
Formula Weight 264.26
Percent Purity ≥96.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Diethyl Phosphate