missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Dicyanamide 97.0+%, TCI America™

Catalog No. p-7129103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06801G 1 g
E06805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06801G Supplier TCI America Supplier No. E06801G

Chemical Identifiers

CAS 370865-89-7
Molecular Formula C8H11N5
Molecular Weight (g/mol) 177.211
InChI Key MKHFCTXNDRMIDR-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h
PubChem CID 11159638
IUPAC Name cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Quantity 1 g
UN Number 2810
Formula Weight 177.21
Percent Purity ≥97.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Dicyanamide