Learn More
1-Ethyl-3-methylimidazolium dicyanamide, 98%
CAS: 370865-89-7 | C8H11N5 | 177.211 g/mol
$218.90 - $1004.45
Chemical Identifiers
| CAS | 370865-89-7 |
|---|---|
| Molecular Formula | C8H11N5 |
| Molecular Weight (g/mol) | 177.211 |
| MDL Number | MFCD08276373 |
| InChI Key | MKHFCTXNDRMIDR-UHFFFAOYSA-N |
| Synonym | 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h |
| PubChem CID | 11159638 |
| IUPAC Name | cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium |
| SMILES | CCN1C=C[N+](=C1)C.C(=[N-])=NC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2690106
|
Thermo Scientific Chemicals
H2690106 |
5 g |
Each for $218.90
|
|
|||||
|
AAH2690118
|
Thermo Scientific Chemicals
H2690118 |
50 g |
Each for $1,004.45
|
|
|||||
Description
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.
Solubility
Soluble in water. Soluble in solvents like methanol, acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 370865-89-7 | |
| 177.211 | |
| MKHFCTXNDRMIDR-UHFFFAOYSA-N | |
| 11159638 | |
| CCN1C=C[N+](=C1)C.C(=[N-])=NC#N |
| C8H11N5 | |
| MFCD08276373 | |
| 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h | |
| cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium |
Specifications
| 370865-89-7 | |
| MFCD08276373 | |
| 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h | |
| MKHFCTXNDRMIDR-UHFFFAOYSA-N | |
| cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium | |
| 11159638 | |
| 98% |
| C8H11N5 | |
| 5 g | |
| Soluble in water. Soluble in solvents like methanol,acetone. | |
| CCN1C=C[N+](=C1)C.C(=[N-])=NC#N | |
| 177.211 | |
| 177.21 | |
| 1-Ethyl-3-methylimidazolium dicyanamide |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only