missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Chloride 98.0+%, TCI America™
$39.14 - $672.92
Chemical Identifiers
| CAS | 65039-09-0 |
|---|---|
| Molecular Formula | C6H11ClN2 |
| Molecular Weight (g/mol) | 146.62 |
| MDL Number | MFCD00074843 |
| InChI Key | BMQZYMYBQZGEEY-UHFFFAOYSA-M |
| Synonym | 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180 |
| PubChem CID | 2734160 |
| ChEBI | CHEBI:61327 |
| IUPAC Name | 1-ethyl-3-methyl-1H-imidazol-3-ium chloride |
| SMILES | [Cl-].CCN1C=C[N+](C)=C1 |
Chemical Identifiers
| 65039-09-0 | |
| 146.62 | |
| BMQZYMYBQZGEEY-UHFFFAOYSA-M | |
| 2734160 | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium chloride |
| C6H11ClN2 | |
| MFCD00074843 | |
| 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180 | |
| CHEBI:61327 | |
| [Cl-].CCN1C=C[N+](C)=C1 |
Specifications
| 65039-09-0 | |
| C6H11ClN2 | |
| 5 g | |
| BMQZYMYBQZGEEY-UHFFFAOYSA-M | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium chloride | |
| 2734160 | |
| 146.62 | |
| Crystalline Powder |
| 84°C | |
| MFCD00074843 | |
| 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180 | |
| [Cl-].CCN1C=C[N+](C)=C1 | |
| 146.62 | |
| CHEBI:61327 | |
| ≥98.0% (HPLC,T) | |
| 1-Ethyl-3-methylimidazolium Chloride [for Molten Salt] |
Safety and Handling
TSCA : Yes