accessibility menu, dialog, popup

UserName

CAS RN 65039-09-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium chloride
Synonyms: 1-ethyl-3-methylimidazolium chloride
Molecular Weight (g/mol): 146.62
Molecular Formula: C6H11ClN2
InChi Key: BMQZYMYBQZGEEY-UHFFFAOYSA-M
SMILES: [Cl-].CCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)