missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Bromide 98.0+%, TCI America™
Supplier: TCI America E054325G
Specifications
| 1-Ethyl-3-methylimidazolium Bromide | |
| 74°C | |
| C6H11BrN2 | |
| 25 g | |
| GWQYPLXGJIXMMV-UHFFFAOYSA-M | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium bromide | |
| 2734235 | |
| ≥98.0% (T) |
| 65039-08-9 | |
| White-Yellow | |
| MFCD03427610 | |
| 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br | |
| [Br-].CCN1C=C[N+](C)=C1 | |
| 191.07 | |
| 191.07 | |
| Crystalline Powder |
Chemical Identifiers
| 65039-08-9 | |
| 191.07 | |
| GWQYPLXGJIXMMV-UHFFFAOYSA-M | |
| 2734235 | |
| [Br-].CCN1C=C[N+](C)=C1 |
| C6H11BrN2 | |
| MFCD03427610 | |
| 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium bromide |
Safety and Handling
TSCA : No