missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Bromide 98.0+%, TCI America™

Catalog No. p-7129056
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05435G 5 g
E054325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05435G Supplier TCI America Supplier No. E05435G
Only null left

Chemical Identifiers

CAS 65039-08-9
Molecular Formula C6H11BrN2
Molecular Weight (g/mol) 191.07
MDL Number MFCD03427610
InChI Key GWQYPLXGJIXMMV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
PubChem CID 2734235
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium bromide
SMILES [Br-].CCN1C=C[N+](C)=C1

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium Bromide