missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid, 97%

Catalog No. AAA1156103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1156103 1 g
AAA1156106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1156103 Supplier Thermo Scientific Chemicals Supplier No. A1156103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50920-65-5 | C7H10N2O2 | 154.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50920-65-5
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00156147
InChI Key VFMGOJUUTAPPDA-UHFFFAOYSA-N
Synonym 1-ethyl-3-methyl-1h-pyrazole-5-carboxylic acid, 1-ethyl-3-methylpyrazole-5-carboxylic acid, 2-ethyl-5-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-ethyl-3-methyl, maybridge1_008579, acmc-20aovb, 1-ethyl-3-methyl-1h-pyrazole-5-carboxylicacid, 1-ethyl-3-methylpyrazole-5-carboxylic, 1h-pyrazole-5-carboxylicacid, 1-ethyl-3-methyl
PubChem CID 2743794
IUPAC Name 2-ethyl-5-methylpyrazole-3-carboxylic acid
SMILES CCN1C(=CC(=N1)C)C(=O)O

Specifications

Melting Point 137°C to 140°C
Quantity 1 g
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only