missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2,3-dioxopiperazine 98.0+%, TCI America™
Supplier: TCI America E0459250G
Specifications
| 1-Ethyl-2,3-dioxopiperazine | |
| 125°C | |
| C6H10N2O2 | |
| 250 g | |
| ZBEKOEYCWKIMGU-UHFFFAOYSA-N | |
| 1-ethylpiperazine-2,3-dione | |
| 108812 | |
| ≥98.0% (GC) |
| 59702-31-7 | |
| White-Yellow | |
| MFCD00051825 | |
| 1-ethyl-2,3-dioxopiperazine, n-ethyl-2,3-dioxopiperazine, 1-ethyl-2,3-piperazinedione, 2,3-piperazinedione, 1-ethyl, 4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-piperazinedione, ccris 8931, 4-ethyl-2,3-dioxo-piperazine, 4-ethyl-2,3-dioxypiperazine, n-ethylpiperazine-2,3-dione | |
| CCN1CCNC(=O)C1=O | |
| 142.158 | |
| 142.16 | |
| Crystalline Powder |
Chemical Identifiers
| 59702-31-7 | |
| 142.158 | |
| ZBEKOEYCWKIMGU-UHFFFAOYSA-N | |
| 108812 | |
| CCN1CCNC(=O)C1=O |
| C6H10N2O2 | |
| MFCD00051825 | |
| 1-ethyl-2,3-dioxopiperazine, n-ethyl-2,3-dioxopiperazine, 1-ethyl-2,3-piperazinedione, 2,3-piperazinedione, 1-ethyl, 4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-piperazinedione, ccris 8931, 4-ethyl-2,3-dioxo-piperazine, 4-ethyl-2,3-dioxypiperazine, n-ethylpiperazine-2,3-dione | |
| 1-ethylpiperazine-2,3-dione |
Safety and Handling
EINECSNumber : (5)-5872&(5)-5806
RTECSNumber : TL6380250
TSCA : No