missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2,3-dioxopiperazine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 125°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 142.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Ethyl-2,3-dioxopiperazine |
Safety and Handling
| EINECSNumber | (5)-5872&(5)-5806 |
| RTECSNumber | TL6380250 |
| TSCA | No |