missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate, 98%, Thermo Scientific™

Catalog No. p-7053283
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5954106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5954106 Supplier Thermo Scientific Chemicals Supplier No. H5954106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

Chemical Identifiers

CAS 307492-75-7
Molecular Formula C7H13BF4N2
Molecular Weight (g/mol) 212.00
MDL Number MFCD08458929
InChI Key KNIWGPCMWBGRBE-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium tetrafluoroborate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium tetrafluoroborate, 3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, dsstox_cid_29122, dsstox_rid_83341, dsstox_gsid_49266, 1-ethyl 2,3 dimethyl imidazolium tetrafluoroborate, 1-ethyl-2,3-dimethylimidazol-1-ium tetrafluoroborate
PubChem CID 21977262
IUPAC Name 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCN1C=C[N+](C)=C1C

Specifications

Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 212
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen