CAS RN 307492-75-7
IUPAC Name:
1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms:
3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol):
212
Molecular Formula:
C7H13BF4N2
InChi Key:
KNIWGPCMWBGRBE-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CCN1C=C[N+](C)=C1C
1
–
1
of
1
results
1
Promotions Available
1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate, 98%, Thermo Scientific™
1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate, 98%, Thermo Scientific™
CAS: 307492-75-7 Molecular Formula: C7H13BF4N2 Molecular Weight (g/mol): 212.00 MDL Number: MFCD08458929 InChI Key: KNIWGPCMWBGRBE-UHFFFAOYSA-N Synonym: 1-ethyl-2,3-dimethylimidazolium tetrafluoroborate,1-ethyl-2,3-dimethyl-1h-imidazol-3-ium tetrafluoroborate,3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate,dsstox_cid_29122,dsstox_rid_83341,dsstox_gsid_49266,1-ethyl 2,3 dimethyl imidazolium tetrafluoroborate,1-ethyl-2,3-dimethylimidazol-1-ium tetrafluoroborate PubChem CID: 21977262 IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide SMILES: F[B-](F)(F)F.CCN1C=C[N+](C)=C1C