missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium chloride, 97%

Catalog No. p-7053165
Change view
Click to view available options
Quantity:
5 g
50 g
Catalog No. Quantity
AAH5922506 5 g
AAH5922518 50 g
2 options

Catalog No. AAH5922506

Supplier: Thermo Scientific Chemicals H5922506

Only null left

CAS: 92507-97-6 | C7H13N2 | 125.19 g/mol

It is used reaction media for chemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used reaction media for chemical processes.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 92507-97-6
Molecular Formula C7H13N2
Molecular Weight (g/mol) 125.19
MDL Number MFCD03093290
InChI Key IRGDPGYNHSIIJJ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium chloride, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride, ksc496m4d, 1,2-dimethyl-3-ethylimidazolium chloride, 1-ethyl-2,3-dimethyl-imidazolium chloride, 1-ethyl-2,3-dimethylimidazol-3-ium chloride, 1-ethyl-2,3-dimethylimidazol-1-ium chloride, 3-ethyl-1,2-dimethylimidazol-1-ium chloride, 1-ethyl-2,3-dimethylimidazolium chloride t
PubChem CID 2734165
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;chloride
SMILES CCN1C=C[N+](C)=C1C

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 160.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium chloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only