CAS RN 92507-97-6
IUPAC Name:
1-ethyl-2,3-dimethyl-1H-imidazol-3-ium
Synonyms:
3-ethyl-1,2-dimethylimidazol-1-ium
Molecular Weight (g/mol):
125.19
Molecular Formula:
C7H13N2
InChi Key:
IRGDPGYNHSIIJJ-UHFFFAOYSA-N
SMILES:
CCN1C=C[N+](C)=C1C
1
–
1
of
1
results
1
Promotions Available
1-Ethyl-2,3-dimethylimidazolium chloride, 97%
1-Ethyl-2,3-dimethylimidazolium chloride, 97%
CAS: 92507-97-6 Molecular Formula: C7H13N2 Molecular Weight (g/mol): 125.19 MDL Number: MFCD03093290 InChI Key: IRGDPGYNHSIIJJ-UHFFFAOYSA-N Synonym: 1-ethyl-2,3-dimethylimidazolium chloride,1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride,ksc496m4d,1,2-dimethyl-3-ethylimidazolium chloride,1-ethyl-2,3-dimethyl-imidazolium chloride,1-ethyl-2,3-dimethylimidazol-3-ium chloride,1-ethyl-2,3-dimethylimidazol-1-ium chloride,3-ethyl-1,2-dimethylimidazol-1-ium chloride,1-ethyl-2,3-dimethylimidazolium chloride t PubChem CID: 2734165 IUPAC Name: 1-ethyl-2,3-dimethylimidazol-3-ium;chloride SMILES: CCN1C=C[N+](C)=C1C