missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7129133
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07535G 5 g
E075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07535G Supplier TCI America Supplier No. E07535G
Only null left

Chemical Identifiers

CAS 174899-90-2
Molecular Formula C9H13F6N3O4S2
Molecular Weight (g/mol) 405.33
InChI Key XDJYSDBSJWNTQT-UHFFFAOYSA-N
PubChem CID 21932259
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-ethyl-2,3-dimethylimidazol-3-ium
SMILES CCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 405.33
Percent Purity ≥98.0% (N,T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium Bis(trifluoromethanesulfonyl)imide