accessibility menu, dialog, popup

UserName

CH3NNH3CH3CFFFSOONSOOFFF

CAS RN 174899-90-2

IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 3-ethyl-1,2-dimethylimidazol-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 405.33
Molecular Formula: C9H13F6N3O4S2
InChi Key: XDJYSDBSJWNTQT-UHFFFAOYSA-N
SMILES: CCN1C=C[N+](C)=C1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results