missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-1-methylpyrrolidinium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. E097725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E097725G 25 g
E09775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E097725G Supplier TCI America Supplier No. E097725G

Chemical Identifiers

CAS 117947-85-0
Molecular Formula C7H16BF4N
Molecular Weight (g/mol) 201.016
MDL Number MFCD08458505
InChI Key LAGDCVJRCOKWTN-UHFFFAOYSA-N
PubChem CID 18374068
IUPAC Name 1-ethyl-1-methylpyrrolidin-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[N+]1(CCCC1)C

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 201.02
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-1-methylpyrrolidinium Tetrafluoroborate