accessibility menu, dialog, popup

UserName

BFFFFCH3NCH3

CAS RN 117947-85-0

IUPAC Name: 1-ethyl-1-methylpyrrolidin-1-ium; tetrafluoroboranuide
Synonyms: 1-ethyl-1-methylpyrrolidin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H16BF4N
InChi Key: LAGDCVJRCOKWTN-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[N+]1(C)CCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results