missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-1-methylpyrrolidinium Bromide 97.0+%, TCI America™
Supplier: TCI America E105025G
Specifications
| 1-Ethyl-1-methylpyrrolidinium Bromide | |
| 307°C | |
| C7H16BrN | |
| 25 g | |
| KHJQQUGSPDBDRM-UHFFFAOYSA-M | |
| 1-ethyl-1-methylpyrrolidin-1-ium;bromide | |
| 112264 | |
| ≥97.0% (T) |
| 69227-51-6 | |
| White-Yellow | |
| MFCD03095384 | |
| 1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d | |
| CC[N+]1(CCCC1)C.[Br-] | |
| 194.116 | |
| 194.12 | |
| Crystalline Powder |
Chemical Identifiers
| 69227-51-6 | |
| 194.116 | |
| KHJQQUGSPDBDRM-UHFFFAOYSA-M | |
| 112264 | |
| CC[N+]1(CCCC1)C.[Br-] |
| C7H16BrN | |
| MFCD03095384 | |
| 1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d | |
| 1-ethyl-1-methylpyrrolidin-1-ium;bromide |
Safety and Handling
EINECSNumber : (5)-6174
TSCA : Yes