missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-1-methylpyrrolidinium Bromide 97.0+%, TCI America™

Catalog No. p-7129303
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10505G 5 g
E105025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10505G Supplier TCI America Supplier No. E10505G
Only null left

Chemical Identifiers

CAS 69227-51-6
Molecular Formula C7H16BrN
Molecular Weight (g/mol) 194.116
MDL Number MFCD03095384
InChI Key KHJQQUGSPDBDRM-UHFFFAOYSA-M
Synonym 1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
PubChem CID 112264
IUPAC Name 1-ethyl-1-methylpyrrolidin-1-ium;bromide
SMILES CC[N+]1(CCCC1)C.[Br-]

Specifications

Melting Point 307°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-1-methylpyrrolidinium Bromide
EINECSNumber (5)-6174
TSCA Yes