missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-1-cyclopentene 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 107°C |
| Quantity | 5 mL |
| UN Number | 3295 |
| Formula Weight | 96.17 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethyl-1-cyclopentene |
Safety and Handling
| TSCA | No |