accessibility menu, dialog, popup

UserName

H3C

CAS RN 2146-38-5

IUPAC Name: 1-ethylcyclopent-1-ene
Synonyms: cyclopentene, 1-ethyl-
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: QYYQTLLGVAPKPN-UHFFFAOYSA-N
SMILES: CCC1=CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results