Learn More
(1-Ethoxycarbonylethylidene)triphenylphosphorane, 97%
CAS: 5717-37-3 | C23H23O2P | 362.41 g/mol
$160.29 - $2,439.66
Chemical Identifiers
| CAS | 5717-37-3 |
|---|---|
| Molecular Formula | C23H23O2P |
| Molecular Weight (g/mol) | 362.41 |
| MDL Number | MFCD00009160 |
| InChI Key | KZENFXVDPUMQOE-UHFFFAOYSA-N |
| Synonym | ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane |
| PubChem CID | 79792 |
| IUPAC Name | ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate |
| SMILES | CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1561909
|
Thermo Scientific Chemicals
A1561909 |
10 g |
N/A
|
|
|||||
|
AAA1561914
|
Thermo Scientific Chemicals
A1561914 |
25 g |
N/A
|
|
|||||
|
AAA1561918
|
Thermo Scientific Chemicals
A1561918 |
50 g |
N/A
|
|
|||||
|
AAA1561930
|
Thermo Scientific Chemicals
A1561930 |
250 g |
N/A
|
|
|||||
Description
(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. Itâs also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. Itâs also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.
Solubility
Soluble in water.
Notes
Air sensitive. Avoid contact with oxygen and air. Store away from oxidizing agent. Contains triphenylphosphine oxide.
Chemical Identifiers
| 5717-37-3 | |
| 362.41 | |
| KZENFXVDPUMQOE-UHFFFAOYSA-N | |
| 79792 | |
| CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C23H23O2P | |
| MFCD00009160 | |
| ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane | |
| ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate |
Specifications
| 5717-37-3 | |
| C23H23O2P | |
| 10 g | |
| Air sensitive | |
| Soluble in water. | |
| CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 362.41 | |
| 362.41 | |
| (1-Ethoxycarbonylethylidene)triphenylphosphorane |
| 154°C to 160°C | |
| MFCD00009160 | |
| 757112 | |
| ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane | |
| KZENFXVDPUMQOE-UHFFFAOYSA-N | |
| ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate | |
| 79792 | |
| 97% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
EINECSNumber : 227-215-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only