missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethylidene)triphenylphosphorane, 97%

Catalog No. AAA1561909
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561909 10 g
AAA1561914 25 g
AAA1561918 50 g
AAA1561930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1561909 Supplier Thermo Scientific Chemicals Supplier No. A1561909

CAS: 5717-37-3 | C23H23O2P | 362.41 g/mol

(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. It’s also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. It’s also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.

Solubility
Soluble in water.

Notes
Air sensitive. Avoid contact with oxygen and air. Store away from oxidizing agent. Contains triphenylphosphine oxide.

Chemical Identifiers

CAS 5717-37-3
Molecular Formula C23H23O2P
Molecular Weight (g/mol) 362.41
MDL Number MFCD00009160
InChI Key KZENFXVDPUMQOE-UHFFFAOYSA-N
Synonym ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane
PubChem CID 79792
IUPAC Name ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate
SMILES CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 154°C to 160°C
Quantity 10 g
Beilstein 757112
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 362.41
Percent Purity 97%
Chemical Name or Material (1-Ethoxycarbonylethylidene)triphenylphosphorane
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 227-215-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only