missing translation for 'onlineSavingsMsg'
Learn More

1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

$270.02 - $867.64

Chemical Identifiers

CAS 864754-16-5
Molecular Formula C13H21BN2O4
Molecular Weight (g/mol) 280.13
MDL Number MFCD08706026
InChI Key YUEZJHOSHBTWPV-UHFFFAOYSA-N
Synonym ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl acetate, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-ethoxycarbonylmethyl pyrazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-yl-acetic acid ethyl ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-acetate, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl acetate, pubchem18443, 1h-pyrazole-1-acetic acid,4-4,4,5,5-tetramethyl-1
PubChem CID 16218385
IUPAC Name ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]acetate
SMILES CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC445010010
View Documents Promotion Details
N/A 1g
Each for $270.02
Only null left
 
AC445010050
View Documents Promotion Details
N/A 5g
Each for $867.64
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

864754-16-5
280.13
YUEZJHOSHBTWPV-UHFFFAOYSA-N
16218385
CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
C13H21BN2O4
MFCD08706026
ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl acetate, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-ethoxycarbonylmethyl pyrazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-yl-acetic acid ethyl ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-acetate, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl acetate, pubchem18443, 1h-pyrazole-1-acetic acid,4-4,4,5,5-tetramethyl-1
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]acetate

Specifications

864754-16-5
96% min. (GC)
C13H21BN2O4
ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl acetate, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-ethoxycarbonylmethyl pyrazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-yl-acetic acid ethyl ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-acetate, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl acetate, pubchem18443, 1h-pyrazole-1-acetic acid,4-4,4,5,5-tetramethyl-1
CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
280.13
280.13
1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester
>110°C
Glass Bottle
MFCD08706026
YUEZJHOSHBTWPV-UHFFFAOYSA-N
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]acetate
16218385
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only