accessibility menu, dialog, popup

UserName

CAS RN 864754-16-5

BOOH3CH3CH3CH3CNNOOCH3

CAS RN 864754-16-5

IUPAC Name: ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetate
Synonyms: ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]acetate
Molecular Weight (g/mol): 280.13
Molecular Formula: C13H21BN2O4
InChi Key: YUEZJHOSHBTWPV-UHFFFAOYSA-N
SMILES: CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)