missing translation for 'onlineSavingsMsg'
Learn More

1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%

Catalog No. AC445010010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445010010 1 g
AC445010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445010010 Supplier Thermo Scientific Chemicals Supplier No. 445010010

CAS: 864754-16-5 | C13H21BN2O4 | 280.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 864754-16-5
Molecular Formula C13H21BN2O4
Molecular Weight (g/mol) 280.13
MDL Number MFCD08706026
InChI Key YUEZJHOSHBTWPV-UHFFFAOYSA-N
Synonym ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl acetate, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 1-ethoxycarbonylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-ethoxycarbonylmethyl pyrazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-yl-acetic acid ethyl ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-acetate, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl acetate, pubchem18443, 1h-pyrazole-1-acetic acid,4-4,4,5,5-tetramethyl-1
PubChem CID 16218385
IUPAC Name ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]acetate
SMILES CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 280.13
Percent Purity 97%
Chemical Name or Material 1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only