missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline 98.0+%, TCI America™

Supplier:  TCI America E036325G

Catalog No. E036325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline
66°C
MFCD00006703
eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
247.29
247.29
Crystalline Powder
16357-59-8
C14H17NO3
25 g
GKQLYSROISKDLL-UHFFFAOYNA-N
ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate
27833
≥98.0% (HPLC,T)

Chemical Identifiers

16357-59-8
247.29
GKQLYSROISKDLL-UHFFFAOYNA-N
27833
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
C14H17NO3
MFCD00006703
eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : VB2010000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions